MAT047027213 Matrix Scientific

Chlorofluoroacetic acid

  • MDL number MFCD00871521
  • Cas Number : 471-44-3
  • Molecular Weight : 112.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.532/25
Storage temp. Ambient
Country of Origin USA
Boiling point 162
Product Link http://www.matrixscientific.com/008872.html

Safety Information

hcodes Corrosive/Irritant
MSDS http://www.matrixscientific.com/008872.html
Availability details for
Quantity
  • Availability
  • Add to Cart