MAT047027218 Matrix Scientific

3-Chloro-2-iodo-1,1,1-trifluoropropane

  • MDL number MFCD01862016
  • Cas Number : 113402-77-0
  • Molecular Weight : 258.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/008876.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008876.html
Availability details for
Quantity
  • Availability
  • Add to Cart