MAT047027224 Matrix Scientific

4'-Chloro-3'-(trifluoromethyl)acetophenone

  • MDL number MFCD03094146
  • Cas Number : 129825-11-2
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-63
Purity 99
Country of Origin USA
Product Link http://www.matrixscientific.com/008882.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008882.html
Availability details for
Quantity
  • Availability
  • Add to Cart