MAT047027269 Matrix Scientific

2,3-Difluoro-4-hydroxybenzonitrile

  • MDL number MFCD00269596
  • Cas Number : 126162-38-7
  • Molecular Weight : 155.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/008922.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008922.html
Availability details for
Quantity
  • Availability
  • Add to Cart