MAT047027635 Matrix Scientific

2-(4-Fluorobenzoyl)-3-(dimethylamino)-acrylonitrile

  • MDL number MFCD03453098
  • Cas Number : 52200-15-4
  • Molecular Weight : 218.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-145
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/008940.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008940.html
Availability details for
Quantity
  • Availability
  • Add to Cart