MAT047027652 Matrix Scientific

Methyl 2-bromo-3,3,3-trifluoropropionate

  • MDL number MFCD04038319
  • Cas Number : 113816-36-7
  • Molecular Weight : 220.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerate
Country of Origin USA
Boiling point 126
Product Link http://www.matrixscientific.com/009025.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/009025.html
Availability details for
Quantity
  • Availability
  • Add to Cart