MAT047027670 Matrix Scientific

5H-Perfluoro-3,4-bis(trifluoromethyl)-hexa-2,4-diene

  • MDL number MFCD06248650
  • Cas Number : 104047-06-5
  • Molecular Weight : 344.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 78-80
Product Link http://www.matrixscientific.com/009089.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009089.html
Availability details for
Quantity
  • Availability
  • Add to Cart