MAT047027680 Matrix Scientific

Perfluoro(2-methyl-3-pentanone)

  • MDL number MFCD04038343
  • Cas Number : 756-13-8
  • Molecular Weight : 316.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -108
Purity 98
Density 1.6
Country of Origin USA
Boiling point 49
Product Link http://www.matrixscientific.com/009117.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009117.html
Availability details for
Quantity
  • Availability
  • Add to Cart