MAT047027683 Matrix Scientific

1-(Perfluoro-n-octyl)dodecane

  • MDL number MFCD04038346
  • Cas Number : 106873-67-0
  • Molecular Weight : 588.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71
Country of Origin USA
Boiling point 479
Product Link http://www.matrixscientific.com/009126.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009126.html
Availability details for
Quantity
  • Availability
  • Add to Cart