MAT047027703 Matrix Scientific

2-(Trifluoromethyl)dioxolane

  • MDL number MFCD04038366
  • Cas Number : 2344-09-4
  • Molecular Weight : 142.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.46
Country of Origin USA
Boiling point 89.5
Product Link http://www.matrixscientific.com/009202.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/009202.html
Availability details for
Quantity
  • Availability
  • Add to Cart