MAT047027704 Matrix Scientific

2-(Trifluoromethyl)-3-ethoxydodecafluorohexane

  • MDL number MFCD04038367
  • Cas Number : 297730-93-9
  • Molecular Weight : 414.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -100°
Purity 99
Density 1.61
Storage temp. Ambient
Country of Origin USA
Boiling point 130°
Product Link http://www.matrixscientific.com/009203.html

Safety Information

MSDS http://www.matrixscientific.com/009203.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart