MAT047027715 Matrix Scientific

1-BOC-4-[4-(Methylsulfonyl)-2-nitrophenyl]-piperidin-4-amine

  • MDL number MFCD04037965
  • Cas Number : 849035-91-2
  • Molecular Weight : 399.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-158
Country of Origin USA
Product Link http://www.matrixscientific.com/009366.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009366.html
Availability details for
Quantity
  • Availability
  • Add to Cart