MAT047027716 Matrix Scientific

N-[4-(Methylsulfonyl)-2-nitrophenyl]-piperidin-4-amine

  • MDL number MFCD06248443
  • Cas Number : 883500-35-4
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-179
Country of Origin USA
Product Link http://www.matrixscientific.com/009368.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009368.html
Availability details for
Quantity
  • Availability
  • Add to Cart