MAT047027720 Matrix Scientific

2-Bromo-4-fluorophenylacetic acid

  • MDL number MFCD04107675
  • Cas Number : 61150-59-2
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-111
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/009436.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009436.html
Availability details for
Quantity
  • Availability
  • Add to Cart