MAT047027970 Matrix Scientific

8-Quinolineboronic acid dimethyl ester

  • MDL number MFCD03788428
  • Cas Number : 1259393-04-8
  • Molecular Weight : 201.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/009806.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/009806.html
Availability details for
Quantity
  • Availability
  • Add to Cart