MAT047027980 Matrix Scientific

2-Amino-5-bromobenzamide

  • MDL number MFCD04037881
  • Cas Number : 16313-66-9
  • Molecular Weight : 215.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-191
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/009823.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009823.html
Availability details for
Quantity
  • Availability
  • Add to Cart