MAT047027990 Matrix Scientific

3-(Aminomethyl)phenol

  • MDL number MFCD00798977
  • Cas Number : 73604-31-6
  • Molecular Weight : 123.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-168
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/009265.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/009265.html
Availability details for
Quantity
  • Availability
  • Add to Cart