MAT047028027 Matrix Scientific

1-(4-Methoxyphenyl)-2-quinolin-2-ylethanone

  • MDL number MFCD04037914
  • Cas Number : 7469-86-5
  • Molecular Weight : 277.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-155
Country of Origin USA
Product Link http://www.matrixscientific.com/009298.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009298.html
Availability details for
Quantity
  • Availability
  • Add to Cart