MAT047028036 Matrix Scientific

2,6-Dichloro-4-methylnicotinamide

  • MDL number MFCD00052636
  • Cas Number : 38841-54-2
  • Molecular Weight : 205.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 170-171
Product Link http://www.matrixscientific.com/009307.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009307.html
Availability details for
Quantity
  • Availability
  • Add to Cart