MAT047028041 Matrix Scientific

1-(2,4-Dimethyl-1,3-oxazol-5-yl)ethanone

  • MDL number MFCD01934451
  • Cas Number : 23012-25-1
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-59
Country of Origin USA
Boiling point 162-164�5 Torr
Product Link http://www.matrixscientific.com/009311.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009311.html
Availability details for
Quantity
  • Availability
  • Add to Cart