MAT047028042 Matrix Scientific

1-(3-Nitropyridin-2-yl)piperidine-4-carbonitrile

  • MDL number MFCD00106383
  • Cas Number : 671191-99-4
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Country of Origin USA
Product Link http://www.matrixscientific.com/009312.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009312.html
Availability details for
Quantity
  • Availability
  • Add to Cart