MAT047028066 Matrix Scientific

4-Fluoro-2-methyl-1-(methylsulfonyl)benzene

  • MDL number MFCD04037935
  • Cas Number : 828270-62-8
  • Molecular Weight : 188.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-105
Country of Origin USA
Product Link http://www.matrixscientific.com/009333.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009333.html
Availability details for
Quantity
  • Availability
  • Add to Cart