MAT047028073 Matrix Scientific

[2-Bromo-4-(methylsulfonyl)phenyl]hydrazine

  • MDL number MFCD04037940
  • Cas Number : 849035-73-0
  • Molecular Weight : 265.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009339.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009339.html
Availability details for
Quantity
  • Availability
  • Add to Cart