MAT047028092 Matrix Scientific

5-Fluoro-7-(methylsulfonyl)-1H-indole-2-carboxylic acid

  • MDL number MFCD04037958
  • Cas Number : 849035-87-6
  • Molecular Weight : 257.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270-272�c)
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/009357.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009357.html
Availability details for
Quantity
  • Availability
  • Add to Cart