MAT047028162 Matrix Scientific

2-tert-Butyl-N-piperidin-4-yl-1H-indol-5-amine

  • MDL number MFCD01319470
  • Cas Number : 397842-07-8
  • Molecular Weight : 271.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-215�c)
Country of Origin USA
Product Link http://www.matrixscientific.com/009419.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009419.html
Availability details for
Quantity
  • Availability
  • Add to Cart