MAT047028167 Matrix Scientific

4-(4-Pyrimidin-4-yl-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD02579383
  • Cas Number : 849924-98-7
  • Molecular Weight : 266.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 365-368�c)
Country of Origin USA
Product Link http://www.matrixscientific.com/009424.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009424.html
Availability details for
Quantity
  • Availability
  • Add to Cart