MAT047028184 Matrix Scientific

Ethyl ethoxymethylene-3-oxo-4,4,4-trifluoro-butyrate

  • MDL number MFCD00052716
  • Cas Number : 571-55-1
  • Molecular Weight : 240.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.239/25
Country of Origin USA
Boiling point 80-82�Torr
Product Link http://www.matrixscientific.com/008932.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008932.html
Availability details for
Quantity
  • Availability
  • Add to Cart