MAT047028250 Matrix Scientific

3-Nitro-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD00274359
  • Cas Number : 116965-16-3
  • Molecular Weight : 235.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/009048.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/009048.html
Availability details for
Quantity
  • Availability
  • Add to Cart