MAT047028316 Matrix Scientific

2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether

  • MDL number MFCD00069095
  • Cas Number : 13838-16-9
  • Molecular Weight : 184.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.517
Storage temp. Ambient
Country of Origin USA
Boiling point 56
Product Link http://www.matrixscientific.com/001888.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001888.html
Availability details for
Quantity
  • Availability
  • Add to Cart