MAT047028355 Matrix Scientific

2H-Perfluoro-5-methyl-3,6-dioxanonane

  • MDL number MFCD00054714
  • Cas Number : 3330-14-1
  • Molecular Weight : 452.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -122
Purity 97
Density 1.659
Country of Origin USA
Boiling point 99�1
Product Link http://www.matrixscientific.com/009115.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009115.html
Availability details for
Quantity
  • Availability
  • Add to Cart