MAT047028359 Matrix Scientific

Perfluorooctanamidine

  • MDL number MFCD00069165
  • Cas Number : 307-31-3
  • Molecular Weight : 412.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/009118.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009118.html
Availability details for
Quantity
  • Availability
  • Add to Cart