MAT047028378 Matrix Scientific

Heptafluorobutyrylamidine

  • MDL number MFCD00236736
  • Cas Number : 375-19-9
  • Molecular Weight : 212.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-51
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/008978.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008978.html
Availability details for
Quantity
  • Availability
  • Add to Cart