MAT047028399 Matrix Scientific

4,4,5,5,6,6,6-Heptafluorohexane-1,2-diol

  • MDL number MFCD03424477
  • Cas Number : 1992-91-2
  • Molecular Weight : 244.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 114�mm
Product Link http://www.matrixscientific.com/008981.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008981.html
Availability details for
Quantity
  • Availability
  • Add to Cart