MAT047028400 Matrix Scientific

4,4,5,5,6,6,6-Heptafluorohexan-1-ol

  • MDL number MFCD00077554
  • Cas Number : 679-02-7
  • Molecular Weight : 228.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 140-141�5mm
Product Link http://www.matrixscientific.com/008982.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008982.html
Availability details for
Quantity
  • Availability
  • Add to Cart