MAT047028414 Matrix Scientific

4,4,5,5,6,6,6-Heptafluorohex-2-en-1-ol

  • MDL number MFCD00155829
  • Cas Number : 37759-88-9
  • Molecular Weight : 226.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.4729
Country of Origin USA
Boiling point 60-65.5�mm
Product Link http://www.matrixscientific.com/008985.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008985.html
Availability details for
Quantity
  • Availability
  • Add to Cart