MAT047028442 Matrix Scientific

1-Fluoro-1,1,2-trichloroethane

  • MDL number MFCD00042303
  • Cas Number : 811-95-0
  • Molecular Weight : 151.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -140
Purity 97
Density 1.4921/20
Storage temp. Ambient
Country of Origin USA
Boiling point 88
Product Link http://www.matrixscientific.com/009178.html

Safety Information

hcodes Ozone Depleter/Irritant-Harmful
MSDS http://www.matrixscientific.com/009178.html
Availability details for
Quantity
  • Availability
  • Add to Cart