MAT047028449 Matrix Scientific

1-Chloro-1,2-dibromo-1,2,2-trifluoroethane

  • MDL number MFCD00039316
  • Cas Number : 354-51-8
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 2.2478/20
Country of Origin USA
Boiling point 93-94
Product Link http://www.matrixscientific.com/001903.html

Safety Information

hcodes Irritant/Moisture Sensitive/Stored under Argon
MSDS http://www.matrixscientific.com/001903.html
Availability details for
Quantity
  • Availability
  • Add to Cart