MAT047028463 Matrix Scientific

1,1,1,3,3,3-Hexafluoroisopropyl2-fluoroacrylate

  • MDL number MFCD03094251
  • Cas Number : 74359-06-1
  • Molecular Weight : 240.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/008992.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/008992.html
Availability details for
Quantity
  • Availability
  • Add to Cart