MAT047028474 Matrix Scientific

Hexafluoro-2-methylisopropyl acrylate

  • MDL number MFCD03094206
  • Cas Number : 53038-41-8
  • Molecular Weight : 236.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 60-61�0mm
Product Link http://www.matrixscientific.com/008993.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/008993.html
Availability details for
Quantity
  • Availability
  • Add to Cart