MAT047028505 Matrix Scientific

2-Acetyl-1-ethylpyrrole

  • MDL number MFCD00191234
  • Cas Number : 39741-41-8
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 82� Torr
Product Link http://www.matrixscientific.com/001913.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001913.html
Availability details for
Quantity
  • Availability
  • Add to Cart