MAT047028507 Matrix Scientific

1,1,2,3,3,3-Hexafluoropropyl difluoromethyl ether

  • MDL number MFCD03094220
  • Cas Number : 56860-85-6
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 47
Product Link http://www.matrixscientific.com/008997.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/008997.html
Availability details for
Quantity
  • Availability
  • Add to Cart