MAT047028543 Matrix Scientific

3-(tert-Butoxymethyl)phenylboronic acid

  • MDL number MFCD03788411
  • Cas Number : 858364-78-0
  • Molecular Weight : 208.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/009764.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/009764.html
Availability details for
Quantity
  • Availability
  • Add to Cart