MAT047028545 Matrix Scientific

Ethyl 3,3,3-trifluoropropionate

  • MDL number MFCD01348113
  • Cas Number : 352-23-8
  • Molecular Weight : 156.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109
Purity 97
Country of Origin USA
Boiling point 50�mm
Product Link http://www.matrixscientific.com/008937.html

Safety Information

hcodes Flammable/Irritant
MSDS http://www.matrixscientific.com/008937.html
Availability details for
Quantity
  • Availability
  • Add to Cart