MAT047028588 Matrix Scientific

Sodium 4-fluorobenzoate

  • MDL number MFCD01661384
  • Cas Number : 499-90-1
  • Molecular Weight : 162.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/009243.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009243.html
Availability details for
Quantity
  • Availability
  • Add to Cart