MAT047028598 Matrix Scientific

3-Fluoro-4-methoxyphenylacetic acid

  • MDL number MFCD00060353
  • Cas Number : 452-14-2
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/009245.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009245.html
Availability details for
Quantity
  • Availability
  • Add to Cart