MAT047028614 Matrix Scientific

Indolin-1-amine hydrochloride

  • MDL number MFCD04037889
  • Cas Number : 92259-86-4
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-203
Country of Origin USA
Product Link http://www.matrixscientific.com/009260.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009260.html
Availability details for
Quantity
  • Availability
  • Add to Cart