MAT047028622 Matrix Scientific

Bis-(1,1-dioxo-tetrahydro-1lambda*6*-thiophen-3-yl)-amine

  • MDL number MFCD00449131
  • Cas Number : 884-51-5
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Country of Origin USA
Product Link http://www.matrixscientific.com/009839.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009839.html
Availability details for
Quantity
  • Availability
  • Add to Cart