MAT047028626 Matrix Scientific

4-Phenyl-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]-pyridine-6-carboxylic acid

  • MDL number MFCD00488388
  • Cas Number : 178456-18-3
  • Molecular Weight : 243.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009843.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009843.html
Availability details for
Quantity
  • Availability
  • Add to Cart