MAT047028671 Matrix Scientific

(4-Methoxy-1,1-dioxo-tetrahydro-1lambda*6*-thiophen-3-ylamino)acetic acid

  • MDL number MFCD00194227
  • Cas Number : 201996-71-6
  • Molecular Weight : 223.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009872.html

Safety Information

MSDS http://www.matrixscientific.com/009872.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart