MAT047028677 Matrix Scientific

4-(3,4-Dimethoxy-phenyl)-3-methyl-isoxazol-5-yl-amine

  • MDL number MFCD00612311
  • Cas Number : 339231-90-2
  • Molecular Weight : 234.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009877.html

Safety Information

MSDS http://www.matrixscientific.com/009877.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart